missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human ENA-78 (CXCL5) (8-78 aa) Recombinant Protein, PeproTech®
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Dimensione della confezione:
100µg
1mg
20µg
250µg
500µg
5µg
Descrizione
Recombinant Human ENA-78 is a 7.8 kDa protein consisting of 71 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
The protein encoded by this gene is an inflammatory chemokine that belongs to the CXC chemokine family. This chemokine is produced concomitantly with interleukin-8 (IL8) in response to stimulation with either interleukin-1 (IL1) or tumor necrosis factor-alpha (TNFA). This chemokine is a potent chemotaxin involved in neutrophil activation.
Specifica
Specifica
| Accession Number | P42830 |
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 6374 |
| Molecular Weight (g/mol) | 7.8 kDa |
| Name | Human ENA-78 (CXCL5) (8-78 aa) |
| Quantity | 100 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto