missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human EN-RAGE DuoSet ELISA

Product Code. 30001770
Change view
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001770 1 Kit (for 5 Plates) Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001770 Supplier R&D Systems™ Supplier No. DY105205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols

Human EN-RAGE DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human EN-RAGE DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human EN-RAGE/S100A12.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit (for 5 Plates)
Species Human
Conjugate Biotin
Binding Target EN-RAGE/S100A12
For Use With (Application) ELISA
Gene ID (Entrez) 6283
Synonym CAAF1, CAAF1Neutrophil S100 protein, CAAFI, CAGC, CAGCS100, CAGCS100 calcium binding protein A12 (calgranulin C), Calcium-binding protein in amniotic fluid 1, Calgranulin C, Calgranulin-C, CGRPEN-RAGE, ENRAGE, Extracellular newly identified RAGE-binding protein, MRP6, p6calgranulin C, protein S100-A12, S100 calcium binding protein A12, S100 calcium-binding protein A12, S100 calcium-binding protein A12 (calgranulin C), S100A12
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.