missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EMP3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4505
This item is not returnable.
View return policy
Description
EMP3 Monoclonal specifically detects EMP3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| EMP3 | |
| Monoclonal | |
| LYOPH | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P54852 | |
| EMP3 | |
| NS0 mouse myeloma cell line transfected with human EMP3 Met1-Glu163 Accession # P54852 | |
| 100 μg | |
| Primary | |
| Detects human EMP. Stains human EMP3 cell transfectants but not irrelevant transfectants. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 439949 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EMP-3, epithelial membrane protein 3, HNMP-1, Protein YMP, YMP, YMPHematopoietic neural membrane protein 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2014 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction