missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EMMPRIN/CD147 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
£180.00 - £1510.00
Specifications
| Antigen | EMMPRIN/CD147 |
|---|---|
| Clone | 109403 |
| Dilution | Western Blot 0.2 ug/mL, ELISA, Knockout Validated |
| Applications | Western Blot, ELISA, Immunoassay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18777789
![]() |
R&D Systems
MAB972-SP |
25 μg |
£180.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18773359
![]() |
R&D Systems
MAB972-100 |
100 μg |
£379.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18783308
![]() |
R&D Systems
MAB972-500 |
500 μg |
£1510.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EMMPRIN/CD147 Monoclonal specifically detects EMMPRIN/CD147 in Human samples. It is validated for Western Blot, ELISA, Knockout Validated.Specifications
| EMMPRIN/CD147 | |
| Western Blot 0.2 ug/mL, ELISA, Knockout Validated | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q54A51 | |
| 682 | |
| Mouse myeloma cell line NS0-derived recombinant human EMMPRIN/CD147 Ala22-His205 Accession # Q54A51 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human EMMPRIN/CD147 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse EMMPRIN is observed. |
| 109403 | |
| Western Blot, ELISA, Immunoassay | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 5F7, basigin, basigin (Ok blood group), BSG, CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
| BSG | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title