missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human EMG1 (aa 4-80) Control Fragment Recombinant Protein

Product Code. 30202565
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30202565

Brand: Invitrogen™ RP104702

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (78%), Rat (78%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65231 (PA5-65231. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

EMG1 involved in 40S ribosomal subunit biogenesis. It seems to play a role a methylation reaction in pre-rRNA processing.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q92979
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10436
Name Human EMG1 (aa 4-80) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 18 S rRNA (pseudouridine(1248)-N1)-methyltransferase; 18 S rRNA (pseudouridine-N1-)-methyltransferase NEP1; 18 S rRNA Psi1248 methyltransferase; C2f; EGK_03301; Emg1; EMG1 N1-specific pseudouridine methyltransferase; EMG1 nucleolar protein homolog; EMG1 nucleolar protein homolog (S. cerevisiae); EMG1, N1-specific pseudouridine methyltransferase; essential for mitotic growth 1; gene rich cluster, C2f; Grcc2f; LOC553478 protein; MGC138092 protein; NEP1; nucleolar essential protein 1; nucleolar protein EMG1 homolog; Nucleolar protein EMG1-like protein; probable ribosome biogenesis protein NEP1; protein C2f; Ribosomal RNA small subunit methyltransferase NEP1; ribosome biogenesis protein NEP1; Unknown (protein for MGC:136360); zgc:136360
Common Name EMG1
Gene Symbol EMG1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PSDGFKPRERSGGEQAQDWDALPPKRPRLGAGNKIGGRRLIVVLEGASLETVKVGKTYELLNCDKHKSILLKNGRDP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.