missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human eIF5A (aa 1-25) Control Fragment Recombinant Protein

Product Code. 30198601
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30198601

Brand: Invitrogen™ RP103395

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (40%), Rat (40%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63944 (PA5-63944. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

mRNA-binding protein involved in translation elongation. Has an important function at the level of mRNA turnover, probably acting downstream of decapping. Involved in actin dynamics and cell cycle progression, mRNA decay and probably in a pathway involved in stress response and maintenance of cell wall integrity. With syntenin SDCBP, functions as a regulator of TP53/p53 and TP53/p53-dependent apoptosis. Regulates also TNF-alpha-mediated apoptosis. Mediates effects of polyamines on neuronal process extension and survival. May play an important role in brain development and function, and in skeletal muscle stem cell differentiation. Also described as a cellular cofactor of human T-cell leukemia virus type I (HTLV-1) Rex protein and of human immunodeficiency virus type 1 (HIV-1) Rev protein, essential for mRNA export of retroviral transcripts.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P63241
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 1984
Name Human eIF5A (aa 1-25) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA410058; D19Wsu54e; Eif4d; eIF-4 D; Eif5a; eIF-5 A; EIF5A1; eIF-5A1; eIF-5 A-1; eIF5AI; eukaryotic initiation factor 5 A; Eukaryotic initiation factor 5 A isoform 1; eukaryotic translation initiation factor 5 A; eukaryotic translation initiation factor 5 A-1; MGC104255; MGC99547; OTTHUMP00000128384; Rev-binding factor; uORF A
Common Name eIF5A
Gene Symbol EIF5A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MCGTGGTDSKTRRPPHRASFLKRLE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.