missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human eIF3i (aa 17-100) Control Fragment Recombinant Protein

Product Code. 30196772
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30196772

Brand: Invitrogen™ RP94035

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56302 (PA5-56302. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13347
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8668
Name Human eIF3i (aa 17-100) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 36 kDa; D4Ertd632e; eIF3 p36; eIF-3-beta; eIF3-beta; Eif3i; eIF3-p36; Eif3s2; Eukaryotic translation initiation factor 3 subunit 2; eukaryotic translation initiation factor 3 subunit I; eukaryotic translation initiation factor 3, subunit 1; eukaryotic translation initiation factor 3, subunit 2 (beta); eukaryotic translation initiation factor 3, subunit 2 (beta, 36 kD); eukaryotic translation initiation factor 3, subunit 2 beta, 36 kDa; eukaryotic translation initiation factor 3, subunit I; predicted protein of HQ2242; PRO2242; subuni; TGF-beta receptor binding protein; TGF-beta receptor-interacting protein 1; TGFbeta receptor-interacting protein 1; TRIP1; TRIP-1
Common Name eIF3i
Gene Symbol Eif3i
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KYNREGDLLFTVAKDPIVNVWYSVNGERLGTYMGHTGAVWCVDADWDTKHVLTGSADNSCRLWDCETGKQLALLKTNSAVRTCG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.