missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human eIF2b beta (aa 36-186) Control Fragment Recombinant Protein

Product Code. 30193829
Click to view available options
Quantity:
100 μL
missing translation for 'unitSize'
100µL
This item is not returnable. View return policy

Product Code. 30193829

missing translation for 'mfr': Invitrogen™ RP103014

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61636 (PA5-61636. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

eIF2b beta is the beta subunit of eukaryotic initiation factor-2B (EIF2B). EIF2B is involved in protein synthesis and exchanges GDP and GTP for its activation and deactivation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49770
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8892
Name Human eIF2b beta (aa 36-186) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA409345; C85417; EIF2B; eIF-2 B GDP-GTP exchange factor subunit beta; Eif2b2; EIF2BB; EIF-2 Bbeta; eukaryotic translation initiation factor 2 B subunit beta; eukaryotic translation initiation factor 2 B, subunit 2 (beta, 39 kD); eukaryotic translation initiation factor 2 B, subunit 2 beta; eukaryotic translation initiation factor 2 B, subunit 2 beta, 39 kDa; S20I15; S20III15; translation initiation factor eIF-2 B subunit beta
Common Name eIF2b beta
Gene Symbol Eif2b2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RETLGLLRQIITDHRWSNAGELMELIRREGRRMTAAQPSETTVGNMVRRVLKIIREEYGRLHGRSDESDQQESLHKLLTSGGLNEDFSFHYAQLQSNIIEAINELLVELEGTMENIAAQALEHIHSNEVIMTIGFSRTVEAFLKEAARKRK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.