missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human eIF2b alpha (aa 219-305) Control Fragment Recombinant Protein

Product Code. 30208823
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30208823

Brand: Invitrogen™ RP104873

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65564 (PA5-65564. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes one of five subunits of eukaryotic translation initiation factor 2B (EIF2B), a GTP exchange factor for eukaryotic initiation factor 2 and an essential regulator for protein synthesis. Mutations in this gene and the genes encoding other EIF2B subunits have been associated with leukoencephalopathy with vanishing white matter.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14232
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 1967
Name Human eIF2b alpha (aa 219-305) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 26 kDa; D5Ertd406e; eIF-2 A; EIF2B; eIF-2 B GDP-GTP exchange factor subunit alpha; Eif2b1; EIF2BA; eIF-2 Balpha; eukaryotic translation initiation factor 2 B subunit alpha; eukaryotic translation initiation factor 2 B, subunit 1 (alpha); eukaryotic translation initiation factor 2 B, subunit 1 (alpha, 26 kDa); eukaryotic translation initiation factor 2 B, subunit 1 alpha; eukaryotic translation initiation factor 2 B, subunit 1 alpha, 26 kDa; GTP-exchange protein; translation initiation factor eIF-2 B subunit alpha
Common Name eIF2b alpha
Gene Symbol EIF2B1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence AKAQNKPFYVVAESFKFVRLFPLNQQDVPDKFKYKADTLKVAQTGQDLKEEHPWVDYTAPSLITLLFTDLGVLTPSAVSDELIKLYL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt