missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EIF2AK4 Partial ORF (NP_001013725, 1401 a.a. - 1500 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00440275-Q01.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16075835

  • £283.00 / 10µg
Estimated Shipment: 08-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

EIF2AK4 belongs to a family of kinases that phosphorylate the alpha subunit of eukaryotic translation initiation factor-2 (EIF2S1; MIM 603907) to downregulate protein synthesis in response to varied cellular stresses (Berlanga et al., 1999 [PubMed 10504407]).[supplied by OMIM]

Sequence: VVSVGQMSMSRAINLTQKLWTAGITAEIMYDWSQSQEELQEYCRHHEITYVALVSDKEGSHVKVKSFEKERQTEKRVLETELVDHVLQKLRTKVTDERNG
Specifications

Specifications

NP_001013725
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VVSVGQMSMSRAINLTQKLWTAGITAEIMYDWSQSQEELQEYCRHHEITYVALVSDKEGSHVKVKSFEKERQTEKRVLETELVDHVLQKLRTKVTDERNG
RUO
EIF2AK4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
440275
EIF2AK4 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GCN2/KIAA1338
EIF2AK4
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers