missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human EID1 (aa 97-183) Control Fragment Recombinant Protein

Product Code. 30181016
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30181016

Brand: Invitrogen™ RP98153

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62323 (PA5-62323. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

E1A-like inhibitor of differentiation-1 (EID-1), an acetyltransferase enzyme, binds both the retinoblastoma protein (Rb), a regulator of cell cycle and tissue specific transcription, and the adenovirus E1A-associated cellular p300 transcriptional co-activator protein. EID-1 inhibits cellular differentiation by blocking the histone acetyltransferase activity of p300. EID-1 also acetylates both histones and non-histone proteins such as NCOA3 co-activator. By acetylating histones, EID-1 gives a specific tag for transcriptional activation. In addition to binding Rb and p300, EID-1 also binds to phosphorylated CREB protein, mediating cAMP gene regulation. EID-1 augments the activity of phosphorylated CREB and activates transcription of cAMP responsive genes as a co-activator.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y6B2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23741
Name Human EID1 (aa 97-183) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 21 kDa pRb-associated protein; 2610002K20Rik; C15orf3; CREBBP/EP300 inhibitor 1; CREBBP/EP300 inhibitory protein 1; CRI1; E1A-like inhibitor of differentiation 1; EID; eid1; EID-1; EP300 interacting inhibitor of differentiation 1; EP300-interacting inhibitor of differentiation 1; IRO45620; NB4 apoptosis related protein; ORF12; PNAS-22; PTD014; Rb- and p300-binding protein EID-1; RBP21; retinoblastoma protein-associated protein; RGD1562702
Common Name EID1
Gene Symbol EID1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DEGEEFDDWEDDYDYPEEEQLSGAGYRVSAALEEADKMFLRTREPALDGGFQMHYEKTPFDQLAFIEELFSLMVVNRLTEELGCDEI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.