missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human EHBP1L1 (aa 649-736) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
missing translation for 'unitSize'
100µL
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (35%), Rat (35%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-140037 (PA5-140037. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Spezifikation
Spezifikation
| Accession Number | Q8N3D4 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 254102 |
| Name | Human EHBP1L1 (aa 649-736) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | EH domain binding protein 1 like 1; EH domain binding protein 1-like 1; EH domain-binding protein 1-like protein 1; EHBP1L1; G430002G23Rik; Tangerin |
| Common Name | EHBP1L1 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur