missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EGR1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF2818
This item is not returnable.
View return policy
Description
EGR1 Polyclonal specifically detects EGR1 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| EGR1 | |
| Polyclonal | |
| Unconjugated | |
| P18146 | |
| EGR1 | |
| E. coli-derived recombinant human EGR1 Gln282-Ser433 Accession # P18146 | |
| 100 μg | |
| Primary | |
| Detects human EGR1 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| AT225Transcription factor ETR103, early growth response 1, early growth response protein 1, EGR-1, G0S30, KROX24, KROX-24, Nerve growth factor-induced protein A, NGFIA, NGFI-ATranscription factor Zif268, TIS8, ZIF-268, Zinc finger protein Krox-24, ZNF225, ZNF225Zinc finger protein 225 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1958 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction