missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EGFR (Research Grade Cetuximab Biosimilar) Antibody, R&D Systems™

Human Monoclonal Antibody
Brand: R&D Systems MAB9577-100
This item is not returnable.
View return policy
Description
EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| EGFR | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human EGFR based on Cetuximab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Cetuximab. This product is for research use only. | |
| Human | |
| Supernatant |
| Flow Cytometry, CyTOF | |
| Hu1 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, EGF R, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ErbB, ErbB1, ERBB1PIG61, HER1, HER-1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
| Human EGF R/ErbB1 | |
| 100 μg | |
| 1956 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction