missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human EGFR F(ab')2 (Research Grade Depatuxizumab Biosimilar) Antibody, Novus Biologicals™
Human Monoclonal Antibody
£218.00 - £500.00
Specifications
| Antigen | EGFR |
|---|---|
| Clone | Hu210 |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18724514
|
Novus Biologicals™
MAB9583-100 |
100 μg |
£500.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18704564
|
Novus Biologicals™
MAB9583-SP |
25 μg |
£218.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EGFR Monoclonal antibody specifically detects EGFR in Human samples. It is validated for ELISASpecifications
| EGFR | |
| ELISA | |
| Unconjugated | |
| Human | |
| Human | |
| avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, EGF R, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ErbB, ErbB1, ERBB1PIG61, HER1, HER-1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
| Human EGFR F(ab')2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| Hu210 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 1956 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze/thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title