missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human EGF R Protein, His Tag
GREENER_CHOICE

Product Code. 15806627
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15806627 1 mg 1mg
15896617 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15806627 Supplier ACROBiosystems Supplier No. EGRH52221MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MALS verified

The epidermal growth factor receptor (EGFR; ErbB-1; HER1 in humans) is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands. The epidermal growth factor receptor is a member of the ErbB family of receptors, a subfamily of four closely related receptor tyrosine kinases: EGFR (ErbB-1), HER2/c-neu (ErbB-2), Her 3 (ErbB-3) and Her 4 (ErbB-4). Mutations affecting EGFR expression or activity could result in cancer.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.5kDa. The protein migrates as 95-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Accession Number NP_005219.2
Molecular Weight (g/mol) 70.5 kDa
Quantity 1 mg
Source Human EGF R, His Tag (EGR-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P00533-1).
Immunogen Leu 25 - Ser 645
Storage Requirements -20°C
Gene Alias EGFR,ERBB,ERBB1,HER1,PIG61,mENA
Common Name EGF R
Conjugate Unconjugated
Species Human
Protein Tag His Tag
Buffer PBS, pH7.4
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype Bio-Markers and CD Antigens
Purity or Quality Grade >95% as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.