missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human EDEM3 (aa 823-931) Control Fragment Recombinant Protein

Product Code. 30194917
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30194917

Brand: Invitrogen™ RP105774

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (72%), Rat (72%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65009 (PA5-65009. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Involved in endoplasmic reticulum-associated degradation (ERAD). Accelerates the glycoprotein ERAD by proteasomes, by catalyzing mannose trimming from Man8GlcNAc2 to Man7GlcNAc2 in the N-glycans. Seems to have alpha 1,2-mannosidase activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BZQ6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 80267
Name Human EDEM3 (aa 823-931) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2310050N11Rik; Alpha-1,2-mannosidase EDEM3; C1orf22; EDEM3; ER degradation enhancer, mannosidase alpha-like 3; ER degradation enhancing alpha-mannosidase like protein 3; ER degradation-enhancing alpha-mannosidase-like 3; ER degradation-enhancing alpha-mannosidase-like protein 3; ER degradation-enhancing -mannosidase-like protein 3; RGD1561496
Common Name EDEM3
Gene Symbol EDEM3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SSQEVDLVDQESSEENSLNSHPESLSLADMDNAASISPSEQTSNPTENHETTNLNGECTDLDNQLQEQSETEEDSNPNVSWGKKVQPIDSILADWNEDIEAFEMMEKDE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.