missing translation for 'onlineSavingsMsg'
Learn More

Human EDA2R/TNFRSF27/XEDAR Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18078901
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18078901 100 μg 100µg
18788727 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18078901 Supplier R&D Systems Supplier No. AF1093

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

EDA2R/TNFRSF27/XEDAR Polyclonal specifically detects EDA2R/TNFRSF27/XEDAR in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDA2R/TNFRSF27/XEDAR
Applications Western Blot, Blockade of Receptor-ligand Interaction
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction
Gene Accession No. Q9HAV5
Gene Alias ectodysplasin A2 receptor, EDA-A2 receptor, EDAA2R, EDA-A2R, EDAA2Rtumor necrosis factor receptor superfamily member XEDAR, EDAR2, TNFRSF27, XEDAR, XEDARTNFRSF27tumor necrosis factor receptor superfamily member 27, X-linked ectodysplasin-A2 receptor
Gene Symbols EDA2R
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human EDA2R/TNFRSF27/XEDAR Met1-Glu136 Accession # Q9HAV5
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60401
Test Specificity Detects human EDA2R/TNFRSF27/XEDAR in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.