missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Ecm 29 (aa 1370-1482) Control Fragment Recombinant Protein

Product Code. 30195822
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195822

Brand: Invitrogen™ RP103820

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84437 (PA5-84437. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Ecm 29 appears to be one of three proteins which associate significantly with proteasomes. Ecm 29 has been shown to enhance the stability of the proteasome. Ecm 29 deletion has been shown to produce 26S proteasomes which dissociated in the absence of ATP. It was thus suggested that Ecm 29 tethers the regulatory complex to the proteasome.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5VYK3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23392
Name Human ECM29 (aa 1370-1482) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI314180; AW558785; BB181316; ECM29; ECM29 homolog, proteasome accessory protein; Ecpas; expressed sequence AI314180; FLJ22036; homolog of yeast Ecm29; KIAA0368; KIAA1962; mKIAA0368; Proteasome adapter and scaffold protein ECM29; Proteasome-associated protein ECM29 homolog; RP11-386D8.2
Common Name ECM29
Gene Symbol ECPAS
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GVGLGTKGGCASVIVSLTTQCPQDLTPYSGKLMSALLSGLTDRNSVIQKSCAFAMGHLVRTSRDSSTEKLLQKLNGWYMEKEEPIYKTSCALTIHAIGRYSPDVLKNHAKEVL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.