missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ECM1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £395.00
Specifications
| Antigen | ECM-1/Secretory Component P85 |
|---|---|
| Clone | 966248 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18767439
![]() |
R&D Systems
MAB3937-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18744859
![]() |
R&D Systems
MAB3937-100 |
100 μg |
£395.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ECM-1/Secretory Component P85 Monoclonal specifically detects ECM-1/Secretory Component P85 in Human samples. It is validated for ELISA.Specifications
| ECM-1/Secretory Component P85 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ECM1, extracellular matrix protein 1, p85, Secretory Component Glycoprotein, Secretory Component P85 | |
| Mouse myeloma cell line NS0-derived recombinant human ECM-1 Ala20-Glu540 Accession # Q16610 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human ECM-1 in direct ELISAs. |
| 966248 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q16610 | |
| 1893 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title