missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ECE2 Full-length ORF (NP_115707.1, 1 a.a. - 255 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16164032
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16164032 10 μg 10µg
16174032 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16164032 Supplier Abnova™ Supplier No. H00009718P01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

Endothelin-converting enzymes, such as ECE2 (EC 3.4.24.71), are type II metalloproteases that generate functionally pleiotropic members of the endothelin vasoactive peptide family (Lorenzo et al., 2001 [PubMed 11718899]).[supplied by OMIM]

Sequence: MASPGAGRAPPELPERNCGYREVEYWDQRYQGAADSAPYDWFGDFSSFRALLEPELRPEDRILVLGCGNSALSYELFLGGFPNVTSVDYSSVVVAAMQARYAHVPQLRWETMDVRKLDFPSASFDVVLEKGTLDALLAGERDPWTVSSEGVHTVDQVLSEVSRVLVPGGRFISMTSAAPHFRTRHYAQAYYGWSLRHATYGSGFHFHLYLMHKGGKLSVAQLALGAQILSPPRPPTSPCFLQDSDHEDFLSAIQL

Specifications

Accession Number NP_115707.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9718
Molecular Weight (g/mol) 54.7kDa
Name ECE2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MASPGAGRAPPELPERNCGYREVEYWDQRYQGAADSAPYDWFGDFSSFRALLEPELRPEDRILVLGCGNSALSYELFLGGFPNVTSVDYSSVVVAAMQARYAHVPQLRWETMDVRKLDFPSASFDVVLEKGTLDALLAGERDPWTVSSEGVHTVDQVLSEVSRVLVPGGRFISMTSAAPHFRTRHYAQAYYGWSLRHATYGSGFHFHLYLMHKGGKLSVAQLALGAQILSPPRPPTSPCFLQDSDHEDFLSAIQL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias KIAA0604/MGC17664/MGC2408/MGC78487
Common Name ECE2
Gene Symbol ECE2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.