missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ECE-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB17842-SP
This item is not returnable.
View return policy
Description
ECE-1 Monoclonal specifically detects ECE-1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| ECE-1 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry | |
| P42892 | |
| ECE1 | |
| Mouse myeloma cell line NS0-derived recombinant human ECE-1 Gln90-Trp770 Accession # P42892 | |
| 25 μg | |
| Primary | |
| Detects human ECE-1 in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 303906 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.24, EC 3.4.24.71, ECE, ECE1, ECE-1, endothelin converting enzyme 1, endothelin-converting enzyme 1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1889 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction