missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EBF3 Partial ORF (NP_001005463, 381 a.a. - 480 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00253738-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16162577

  • £425.00 / 25µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: LYGMPHNNQEIILKRAADIAEALYSVPRNHNQIPTLGNNPAHTGMMGVNSFSSQLAVNVSETSQANDQVGYSRNTSSVSPRGYVPSSTPQQSNYNTVSTS
Specifications

Specifications

NP_001005463
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LYGMPHNNQEIILKRAADIAEALYSVPRNHNQIPTLGNNPAHTGMMGVNSFSSQLAVNVSETSQANDQVGYSRNTSSVSPRGYVPSSTPQQSNYNTVSTS
RUO
EBF3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
253738
EBF3 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
COE3/O/E-2
EBF3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers