missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human EAPP (aa 32-142) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51774 (PA5-51774. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This protein may play an important role in the fine-tuning of both major E2F1 activities, the regulation of the cell-cycle and the induction of apoptosis. Promotes S-phase entry, and inhibits p14(ARP) expression. The E2F family plays a crucial role in the control of cell cycle and action of tumor suppressor proteins and is also a target of the transforming proteins of small DNA tumor viruses. The E2F proteins contain several evolutionally conserved domains found in most members of the family. These domains include a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. The C-terminal of EAPP binds the N-terminal of E2F1 and also interacts with E2F2 and E2F3, but not E2F4.
Specifications
Specifications
| Accession Number | Q56P03 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 55837 |
| Name | Human EAPP (aa 32-142) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1810011O16Rik; BM036; BM-036; c14orf11; E2F associated phosphoprotein; E2F-associated phosphoprotein; Eapp; hypothetical protein LOC327318; RGD1309624; Unknown (protein for MGC:139672); wu:fd21g02; zgc:65820 |
| Common Name | EAPP |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction