missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human E-selectin (CD62E) Recombinant Protein

Product Code. 11544231
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11544231 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11544231 Supplier Invitrogen™ Supplier No. RP75540

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This product contains purified recombinant human e-Selectin protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.

Leukocytes adhere to the blood vessel endothelium during extravasation in postcapillary venules of lymph nodes. In addition, leukocyte adhesion occurs in the capillaries and small venules at any site in the body after onset of inflammation. This response is immediate and involves a cascade of adhesion receptors. At the endothelial surface members of the selectin and immunoglobulin superfamilies participate in this cascade. Selectins are C-type cell surface lectins that play a role in leukocyte adhesion to the blood vessel wall endothelium. E-selectin (CD62E) is an endothelial cell specific selectin that is expressed only after activation with proinflammatory cytokines. In vitro experiments have shown that IL-1, TNFalpha and bacterial wall components like lipopolysaccharides induce the transcription of E-selectin in a NFkB dependent signaling cascade. E-selectin has been associated with blood vessel endothelium in diverse inflammatory situations.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P16581
Concentration 0.105 mg/mL
For Use With (Application) Control, Western Blot
Formulation 0.02 M tris HCl with 100 mM NaCl, 1 mM EDTA, 50% glycerol and no preservative; pH 8
Gene ID (Entrez) 6401
Name Human E-selectin (CD62E)
Quantity 100 μg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias CD62 antigen-like family member E; CD62E; e selectin; Elam; ELAM1; ELAM-1; endothelial adhesion molecule 1; endothelial leukocyte adhesion molecule; endothelial leukocyte adhesion molecule 1; endothelial leukocyte adhesion molecule I; ESEL; E-selectin; LECAM2; leukocyte endothelial cell adhesion molecule 2; leukocyte-endothelial cell adhesion molecule 2; RP1-117P20.2; SELE; selectin E; selectin E (endothelial adhesion molecule 1); selectin, endothelial cell
Common Name E-selectin (CD62E)
Gene Symbol SELE
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Asn179- Leu557 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.