missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human E-Cadherin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 11 publications
Brand: R&D Systems MAB1838-500
This item is not returnable.
View return policy
Description
E-Cadherin Monoclonal specifically detects E-Cadherin in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| E-Cadherin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 2-25 ug/mL | |
| P12830 | |
| CDH1 | |
| Mouse myeloma cell line NS0-derived recombinant human E-Cadherin Asp155-Ile707 Accession # P12830 | |
| 500 μg | |
| Primary | |
| Detects human E-Cadherin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Cadherin-8, rhCadherin-17, recombinant mouse E-Cadherin, rhN-Cadherin, rhP-Cadherin, or rhVE-Cadherin is observed. | |
| Human | |
| Supernatant |
| Western Blot, Immunohistochemistry | |
| 180215 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Arc-1, CAD1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDH1, CDHE, cell-CAM 120/80, Cell-CAM120/80, ECAD, E-cadherin, ECadherin, Epithelial cadherin, LCAM, L-CAM, UVOE-Cadherin, uvomorulin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 999 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction