missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human E-Cadherin Alexa Fluor 647-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems FAB748R-025
This item is not returnable.
View return policy
Description
E-Cadherin Polyclonal specifically detects E-Cadherin in Human samples. It is validated for Flow Cytometry.
Specifications
| E-Cadherin | |
| Polyclonal | |
| Flow Cytometry 5 uL/10^6 cells | |
| P12830 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 999 | |
| Human | |
| IgG |
| Flow Cytometry | |
| Alexa Fluor 647 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Arc-1, CAD1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDH1, CDHE, cell-CAM 120/80, Cell-CAM120/80, ECAD, E-cadherin, ECadherin, Epithelial cadherin, LCAM, L-CAM, UVOE-Cadherin, uvomorulin | |
| Mouse myeloma cell line NS0-derived recombinant human E-Cadherin Asp155-Ile707 Accession # P12830 | |
| 25 Tests | |
| Primary | |
| Detects human E-Cadherin in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction