missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Dynactin 4 (aa 352-443) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58214 (PA5-58214. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May play an role in the regulation of Ins(1,4,5)P3 around the endoplasmic reticulum. Target protein's p62 subunit of the dynactin complex involved in the cytoplasmic dynein motor machinery.
Specifications
Specifications
| Accession Number | Q9UJW0 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 51164 |
| Name | Human Dynactin 4 (aa 352-443) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1110001K06Rik; 4930547K17Rik; C130039E17Rik; DCTN4; Dyn4; dynactin 4; dynactin 4 (p62); dynactin 4 (p62) isoform 1; dynactin 4 (p62) isoform 2; dynactin p62 subunit; dynactin subunit 4; Dynactin subunit p62; hypothetical protein LOC550479; P62; si:ch211-174h24.2; zgc:112431 |
| Common Name | Dynactin 4 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction