missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DUSP27 (aa 250-282) Control Fragment Recombinant Protein

Product Code. 30204883
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30204883

Brand: Invitrogen™ RP90298

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64945 (PA5-64945. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

DUSP27 (Dual Specificity Phosphatase 27, Atypical) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include phosphatase activity and protein tyrosine/serine/threonine phosphatase activity. An important paralog of this gene is DUSP13.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5VZP5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 92235
Name Human DUSP27 (aa 250-282) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias atypical dual-specific protein phosphatase; dual specificity phosphatase 27; dual specificity phosphatase 27 (putative); dual specificity phosphatase and pro isomerase domain containing 1; dual specificity phosphatase DUPD1; DUPD1; DUSP27; FMDSP; Inactive dual specificity phosphatase 27
Common Name DUSP27
Gene Symbol DUSP27
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EALMTVRKKRAIYPNEGFLKQLRELNEKLMEER
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.