missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DSPG3 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB3110
This item is not returnable.
View return policy
Description
DSPG3/EPYC Monoclonal specifically detects DSPG3/EPYC in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| DSPG3/EPYC | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q99645 | |
| EPYC | |
| Mouse myeloma cell line NS0-derived recombinant human DSPG3 Ala20-Val322 Accession # Q99645 | |
| 100 μg | |
| Primary | |
| Detects human DSPG3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse DSPG3 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 329707 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Dermatan sulfate proteoglycan 3dermatan sulphate proteoglycan 3, DSPG3proteoglycan-lb, epiphycan, EPYC, PG-Lb, PGLB, Proteoglycan-Lb, SLRR3B, SLRR3Bepiphycan proteoglycan, Small chondroitin/dermatan sulfate proteoglycan | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1833 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction