missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DSPG3 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3110
This item is not returnable.
View return policy
Description
DSPG3/EPYC Polyclonal specifically detects DSPG3/EPYC in Human samples. It is validated for Western Blot.
Specifications
| DSPG3/EPYC | |
| Polyclonal | |
| Unconjugated | |
| Q99645 | |
| EPYC | |
| Mouse myeloma cell line NS0-derived recombinant human DSPG3 Met1-Val322 Accession # Q99645 | |
| 100 μg | |
| Primary | |
| Detects human DSPG3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant mouse DSPG3 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Dermatan sulfate proteoglycan 3dermatan sulphate proteoglycan 3, DSPG3proteoglycan-lb, epiphycan, EPYC, PG-Lb, PGLB, Proteoglycan-Lb, SLRR3B, SLRR3Bepiphycan proteoglycan, Small chondroitin/dermatan sulfate proteoglycan | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1833 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction