missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DSCAM Long Isoform Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB36661-SP
This item is not returnable.
View return policy
Description
DSCAM Monoclonal specifically detects DSCAM in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| DSCAM | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| O60469 | |
| DSCAM | |
| Mouse myeloma cell line NS0-derived recombinant human DSCAM Long Isoform Glu18-Met1595 Accession # O60469 | |
| 25 μg | |
| Primary | |
| Detects human DSCAM Long Isoform in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human DSCAM-L1 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 399212 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CHD2-42, CHD2-52, Down syndrome cell adhesion molecule, human CHD2-52 down syndrome cell adhesion molecule, 10CHD2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1826 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction