missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DSCAM-L1/DSCAM2 ELISA Kit
Human DSCAM-L1/DSCAM2 quantitates human DSCAM-L1/DSCAM2 in serum, plasma, supernatant.
£540.00 - £3870.00
Specifications
Accession Number | 4921507G06Rik; 4930435C18Rik; BB239540; Down syndrome cell adhesion molecule 2; Down syndrome cell adhesion molecule like 1; Down syndrome cell adhesion molecule-like 1; Down syndrome cell adhesion molecule-like protein 1; Down syndrome cell adhesion molecule-like protein 1 homolog; Downs syndrome cell adhesion molecule like 1; DS cell adhesion molecule like 1; DSCAM2; DSCAML1; DSCAM-like 1; KIAA1132; mKIAA1132 |
---|---|
Assay Range | 0.82 to 200 ng/mL |
Assay Sensitivity | 0.82 ng/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17114919
|
Invitrogen™
EH164RBX5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
17824080
|
Invitrogen™
EH164RBX10 |
10 x 96 Tests |
£3870.00
Pack of 10 |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
17164443
|
Invitrogen™
EH164RB |
96 Tests |
£540.00
Pack of 96 |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
Description
Human DSCAM-L1/DSCAM2 quantitates human DSCAM-L1/DSCAM2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human DSCAM-L1/DSCAM2.
DSCAML1 encodes a protein that is a member of the Ig superfamily of cell adhesion molecules and is involved in neuronal differentiation. The encoded membrane-bound protein localizes to the cell surface, where it forms aggregates that repel neuronal processes of the same cell type.Specifications
4921507G06Rik; 4930435C18Rik; BB239540; Down syndrome cell adhesion molecule 2; Down syndrome cell adhesion molecule like 1; Down syndrome cell adhesion molecule-like 1; Down syndrome cell adhesion molecule-like protein 1; Down syndrome cell adhesion molecule-like protein 1 homolog; Downs syndrome cell adhesion molecule like 1; DS cell adhesion molecule like 1; DSCAM2; DSCAML1; DSCAM-like 1; KIAA1132; mKIAA1132 | |
0.82 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
DSCAML1 | |
<10% | |
HRP | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL | |
DSCAML1 | |
4 hr 45 min |
0.82 to 200 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q8TD84 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |