missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DSCAM-L1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3315-SP
This item is not returnable.
View return policy
Description
DSCAM-L1 Polyclonal specifically detects DSCAM-L1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| DSCAM-L1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Down syndrome cell adhesion molecule like 1, Down syndrome cell adhesion molecule-like protein 1, Downs syndrome cell adhesion molecule like 1, DSCAM2, DSCAML1, KIAA1132Down syndrome cell adhesion molecule 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 57453 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q8TD84 | |
| DSCAML1 | |
| Mouse myeloma cell line NS0-derived recombinant human DSCAM-L1 Glu19-Lys1591 Accession # Q8TD84 | |
| 25 μg | |
| Primary | |
| Detects human DSCAM-L1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant human DSCAM is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction