missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DRIL1 (aa 354-480) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52106 (PA5-52106. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The mouse Bright (B-cell regulator of IgH transcription) protein is expressed specifically in B cells and transactivates an IgH enhancer in transient transfection assays. Bright and the Drosophila 'dead ringer' (dri) gene product belong to a protein family whose members share a conserved DNA-binding domain termed the 'A/T-rich interaction domain' (ARID).
Specifications
Specifications
| Accession Number | Q99856 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 1820 |
| Name | Human DRIL1 (aa 354-480) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | ARID domain-containing 3 A; ARID domain-containing protein 3 A; ARID3A; AT rich interactive domain 3 A (Bright like); AT rich interactive domain 3 A (BRIGHT- like) protein; AT rich interactive domain 3 A (BRIGHT-like); AT-rich interaction domain 3 A; AT-rich interactive domain-containing protein 3 A; B-cell regulator of IgH transcription; BRIGHT; dead ringer homolog 1; dead ringer-like 1; dead ringer-like protein 1; Dri1; DRIL1; DRIL3; DRX; E2F-binding protein 1; E2FBP1 |
| Common Name | DRIL1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction