missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Drebrin 1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7739
This item is not returnable.
View return policy
Description
Drebrin 1 Monoclonal specifically detects Drebrin 1 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Specifications
| Drebrin 1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q16643 | |
| DBN1 | |
| E. coli-derived recombinant human Drebrin 1 Asn482-Asp649 (Ser553Pro) Accession # Q16643 | |
| 100 μg | |
| Primary | |
| Detects human Drebrin 1 in ELISAs. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 838102 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| D0S117E, DBN1, Developmentally-regulated brain protein, DKFZp434D064, Drba, drebrin, drebrin 1, Drebrin A, drebrin E, Drebrin E2, drebrin-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1627 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction