missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DQX1 (aa 182-279) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (76%), Rat (76%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58674 (PA5-58674. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
DQX1 (DEAQ box RNA-dependent ATPase 1), also known as FLJ23757, is a 71 amino acid protein that contains one helicase ATP-binding domain and one helicase C-terminal domain. Localized to the nucleus, DQX1 catalyzes the conversion of ATP to ADP and a phosphate. Expressed as three isoforms produced by alternative splicing events, DQX1 is encoded by a gene that maps to human chomosome 2. As the second largest human chromosome, chromosome 2 makes up approximately 8% of the human genome and contains 237 million bases encoding over 1,400 genes. A number of genetic diseases are linked to genes on chromosome 2, including Harlequin icthyosis, sitosterolemia and Alstrom syndrome.
Specifications
Specifications
| Accession Number | Q8TE96 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 165545 |
| Name | Human DQX1 (aa 182-279) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 2310066E11Rik; ATP-dependent RNA helicase DQX1; DEAQ box polypeptide 1; DEAQ box polypeptide 1 (RNA-dependent ATPase); DEAQ box RNA-dependent ATPase 1; DEAQ RNA-dependent ATPase; DEAQ RNA-dependent ATPase DQX1; DEAQ-box RNA dependent ATPase 1; Dqx1 |
| Common Name | DQX1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction