missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DPPII/QPP/DPP7 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB34381-SP
This item is not returnable.
View return policy
Description
DPPII/QPP/DPP7 Monoclonal specifically detects DPPII/QPP/DPP7 in Human samples. It is validated for Western Blot.
Specifications
| DPPII/QPP/DPP7 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| AAH11907 | |
| DPP7 | |
| Mouse myeloma cell line NS0-derived recombinant human DPPII/QPP/DPP7 Gly22-Leu492 Accession # AAH11907 | |
| 25 μg | |
| Primary | |
| Detects human DPPII/QPP/DPP7 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 5% cross-reactivity with recombinant mouse DPP7 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 398024 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| carboxytripeptidase, Dipeptidyl aminopeptidase II, dipeptidyl arylamidase II, dipeptidyl peptidase 2, Dipeptidyl peptidase 7, Dipeptidyl peptidase II, dipeptidylpeptidase 7, dipeptidyl-peptidase 7, dipeptidyl-peptidase II, DPP II, DPP2, DPP7, DPPII, EC 3.4.14.2, QPP, Quiescent cell proline dipeptidase | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 29952 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction