missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DPPA5/ESG1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB3125
Additional Details : Weight : 0.00970kg
Description
DPPA5/ESG1 Monoclonal specifically detects DPPA5/ESG1 in Human samples. It is validated for Western Blot.Specifications
DPPA5/ESG1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
A6NC42 | |
DPPA5 | |
E. coli-derived recombinant human DPPA5/ESG1 Met1-Lys116 Accession # A6NC42 | |
100 μg | |
Primary | |
Detects human DPPA5/ESG1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human DPPA4 or recombinant mouse DPPA5. | |
Human | |
Purified |
Western Blot | |
319606 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
developmental pluripotency associated 5, developmental pluripotency-associated 5 protein, embryonal stem cell specific gene 1, Embryonal stem cell-specific gene 1 protein, Esg1, ESG1ESG-1, hDPPA5 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
340168 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
For Research Use Only