missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DPP9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5419
This item is not returnable.
View return policy
Description
DPP9 Monoclonal specifically detects DPP9 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Specifications
| DPP9 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q1ZZB8 | |
| DPP9 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human DPP9 Arg2-Leu892 (predicted) Accession # Q1ZZB8 | |
| 100 μg | |
| Primary | |
| Detects human DPP9 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) DPPIV/CD26 or rhDPP10 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 757004 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| dipeptidyl peptidase 9, Dipeptidyl peptidase IV-related protein 2, dipeptidyl peptidase IV-related protein-2, Dipeptidyl peptidase IX, Dipeptidyl peptidase-like protein 9, dipeptidylpeptidase 9, dipeptidyl-peptidase 9, DKFZp762F117, DP9, DPLP9, DPP IX, DPRP2, DPRP-2, EC 3.4.14.5, FLJ16073 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 91039 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction