missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DPP6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB23601-SP
This item is not returnable.
View return policy
Description
DPP6 Monoclonal specifically detects DPP6 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| DPP6 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P42658 | |
| DPP6 | |
| Mouse myeloma cell line NS0-derived recombinant human DPP6 Thr118-Asp865 Accession # P42658 | |
| 25 μg | |
| Primary | |
| Detects human DPP6 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human DPPIV/CD26 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 274311 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| dipeptidyl aminopeptidase IV-related protein, dipeptidyl aminopeptidase-like protein 6, Dipeptidyl aminopeptidase-related protein, Dipeptidyl peptidase 6, Dipeptidyl peptidase IV-like protein, dipeptidyl peptidase IV-related protein, Dipeptidyl peptidase VI, dipeptidylpeptidase 6, dipeptidyl-peptidase 6, dipeptidylpeptidase VI, DPP VI, DPPX, DPPXFLJ55680, MGC46605, VF2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1804 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction