missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DPP6 (aa 582-662) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62201 (PA5-62201. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
DPP6 encodes a single-pass type II membrane protein that is a member of the S9B family in clan SC of the serine proteases. This protein has no detectable protease activity, most likely due to the absence of the conserved serine residue normally present in the catalytic domain of serine proteases. However, it does bind specific voltage-gated potassium channels and alters their expression and biophysical properties. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Specifications
| Accession Number | P42658 |
| Concentration | 2.10 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 1804 |
| Name | Human DPP6 (aa 582-662) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | B930011P16Rik; D5Buc3; D5Buc4; D5Buc5; dipeptidyl aminopeptidase IV-related protein; dipeptidyl aminopeptidase-like protein 6; Dipeptidyl aminopeptidase-related protein; Dipeptidyl peptidase 6; Dipeptidyl peptidase IV-like protein; dipeptidyl peptidase IV-related protein; dipeptidyl peptidase like 6; dipeptidyl peptidase VI; dipeptidylpeptidase 6; dipeptidyl-peptidase 6; dipeptidylpeptidase VI; DPL1; DPP VI; Dpp6; Dpp-6; DPPX; Gm1377; In(5)6 H p; In(5)6 H-p; inversion, Chr 5, Harwell 6, proximal; MRD33; Peplb; Rw; VF2 |
| Common Name | DPP6 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction