missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DPP10 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4100-SP
This item is not returnable.
View return policy
Description
DPP10 Polyclonal specifically detects DPP10 in Human samples. It is validated for Western Blot.
Specifications
| DPP10 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| dipeptidyl peptidase 10, Dipeptidyl peptidase IV-related protein 3, dipeptidyl peptidase like protein 2, Dipeptidyl peptidase X, Dipeptidyl peptidase-like protein 2, dipeptidyl-peptidase 10, dipeptidyl-peptidase 10 (non-functional), DPL2, DPP X, DPPY, DPRP3, DPRP-3, DPRP3dipeptidylpeptidase 10, inactive dipeptidyl peptidase 10, KIAA1492 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 57628 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| NP_065919 | |
| DPP10 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human DPP10 Leu56-Glu796 (Pro340Ala) Accession # NP_065919 | |
| 25 μg | |
| Primary | |
| Detects human DPP10 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) DPP6 and rhDPP4 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction