missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DPCR-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3347-SP
This item is not returnable.
View return policy
Description
DPCR-1 Monoclonal specifically detects DPCR-1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| DPCR-1 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| AAH69477 | |
| DPCR1 | |
| NS0 mouse myeloma cell line transfected with human DPCR-1 Met1-Arg235 Accession # AAH69477 | |
| 25 μg | |
| Primary | |
| Detects human human DPCR-1. Stains human DPCR-1-transfected cells but not irrelevant transfectants. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 213201 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| bCX105N19.6, C6orf37, diffuse panbronchiolitis critical region 1, DPCR1, MGC126710, MGC126712, PBLT, PBLTdiffuse panbronchiolitis critical region 1 protein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 135656 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction