missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Dopaminergic Neuron Immunocytochemistry Kit

Product Code. 15994194
Change view
Click to view available options
Quantity:
40 reactions
Unit Size:
40 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15994194 40 reactions 40 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15994194 Supplier Invitrogen™ Supplier No. A29515
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Human Dopaminergic Neuron Immunocytochemistry Kit enables optimal image-based analysis of three key markers of neural differentiation: OTX2, FoxA2, and tyrosine hydroxylase (TH).

This high-performance immunocytochemistry (ICC) kit includes a complete set of primary and secondary antibodies, a nuclear DNA stain, and premade buffers for an optimized staining experiment.

The Human Dopaminergic Neuron Immunocytochemistry Kit enables you to:
• Confirm expression of key cellular markers with highly specific antibodies
• Get more information per sample by measuring more than one marker at a time
• Generate beautiful multiplexed images with minimal effort
• Save time and avoid cell loss by minimizing unnecessary wash steps in your staining protocol
• Process samples with confidence using a complete and optimized set of ICC reagents

Confirm expression of key cellular markers
The antibodies included in the kit have been carefully selected and validated for high performance in the ICC analysis of floor plate progenitors and dopaminergic neurons. OTX2 and FoxA2 are critical nuclear markers while TH is an important cytoplasmic marker.

Get more information per sample
The antibody combinations in the kit have been designed to allow for the specific and simultaneous assessment of two markers at a time (e.g., OTX2 and FoxA2, and FoxA2 and TH) along with nuclear DNA staining to help save time and precious sample.

Generate beautiful multiplexed images
Powered by the superior performance of bright, photostable Alexa Fluor™ dyes, this kit enables the acquisition of beautiful multiplexed images of key neural differentiation marker expression using commonly available blue, green, and orange/red filter sets.

Minimize unnecessary wash steps
The streamlined method recommended for this kit eliminates unnecessary wash steps found in standard ICC protocols, helping to save time and decrease the likelihood of cell loss during the staining protocol. The only washes required are after incubation with primary and secondary antibodies.

Process samples with confidence
An optimized set of premade fixation, permeabilization, blocking, and wash buffers allows you to focus on answering scientific questions rather than spending time preparing reagents and determining compatibility.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 40 reactions
Content And Storage
  • 10 μL of Goat Anti-Human OT x 2 (2000 X)
  • 10 μL of Mouse Anti-Human FoxA2 (1000 X)
  • 10 μL of Rabbit Anti-Human Tyrosine Hydroxylase (1000 X)
  • 20 μL of Alexa Fluor 488 Donkey Anti-Goat IgG (250 X)
    • 20 μL of Alexa Fluor 488 Donkey Anti-Rabbit IgG (250 X)
    • 20 μL of Alexa Fluor 555 Donkey Anti-Mouse (250 X)
    • 20 μL of Alexa Fluor 594 Donkey Anti-Mouse (250 X)
    • 1 vial of NucBlue Fixed Cell ReadyProbes Reagent (1 X)
    • 10 mL of Fixative Solution (1 X)
    • 10 mL of Permeabilization Solution (1 X)
    • 20 mL of Blocking Solution (1 X)
    • 20 mL of Wash Buffer (10 X)

      Store at -20°C to +4°C
Shipping Condition Dry Ice
Product Type Immunocytochemistry Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.