missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DONSON (aa 450-554) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (74%), Rat (74%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61865 (PA5-61865. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
DONSON is essential for DNA amplification in the ovary and required for cell proliferation during development.
Spécification
Spécification
| Accession Number | Q9NYP3 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 29980 |
| Name | Human DONSON (aa 450-554) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | B17; C21orf60; C2TA; DONSON; downstream neighbor of SON; protein downstream neighbor of Son |
| Common Name | DONSON |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu