missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DOM3Z Full-length ORF (NP_005501.2, 1 a.a. - 396 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16184151
Change view
Click to view available options
Quantity:
10 ug
25 ug
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16184151 25 ug 25µg
16174151 10 ug 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16184151 Supplier Abnova™ Supplier No. H00001797P01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6. The function of its protein product is unknown, but its ubiquitous expression and conservation in both simple and complex eukaryotes suggests that this may be a housekeeping gene. [provided by RefSeq]

Sequence: MDPRGTKRGAEKTEVAEPRNKLPRPAPSLPTDPALYSGPFPFYRRPSELGCFSLDAQRQYHGDARALRYYSPPPTNGPGPNFDLRDGYPDRYQPRDEEVQERLDHLLCWLLEHRGRLEGGPGWLAEAIVTWRGHLTKLLTTPYERQEGWQLAASRFQGTLYLSEVETPNARAQRLARPPLLRELMYMGYKFEQYMCADKPGSSPDPSGEVNTNVAFCSVLRSRLGSHPLLFSGEVDCTDPQAPSTQPPTCYVELKTSKEMHSPGQWRSFYRHKLLKWWAQSFLPGVPNVVAGFRNPDGFVSSLKTFPTMKMFEYVRNDRDGWNPSVCMNFCAAFLSFAQSTVVQDDPRLVHLFSWEPGGPVTVSVHQDAPYAFLPIWYVEAMTQDLPSPPKTPSPK

Specifications

Accession Number NP_005501.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 1797
Molecular Weight (g/mol) 71.3kDa
Name DOM3Z (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MDPRGTKRGAEKTEVAEPRNKLPRPAPSLPTDPALYSGPFPFYRRPSELGCFSLDAQRQYHGDARALRYYSPPPTNGPGPNFDLRDGYPDRYQPRDEEVQERLDHLLCWLLEHRGRLEGGPGWLAEAIVTWRGHLTKLLTTPYERQEGWQLAASRFQGTLYLSEVETPNARAQRLARPPLLRELMYMGYKFEQYMCADKPGSSPDPSGEVNTNVAFCSVLRSRLGSHPLLFSGEVDCTDPQAPSTQPPTCYVELKTSKEMHSPGQWRSFYRHKLLKWWAQSFLPGVPNVVAGFRNPDGFVSSLKTFPTMKMFEYVRNDRDGWNPSVCMNFCAAFLSFAQSTVVQDDPRLVHLFSWEPGGPVTVSVHQDAPYAFLPIWYVEAMTQDLPSPPKTPSPK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DOM3L/NG6
Common Name DOM3Z
Gene Symbol DOM3Z
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.