missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DOK6 (aa 232-269) Control Fragment Recombinant Protein

Product Code. 30202382
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30202382

Brand: Invitrogen™ RP107246

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

DOK-6 (docking protein 6), is a novel member of the Dok-4/5 subclass of the p62 Dok family of intracellular adaptor molecules. Dok-6 is highly expressed in the developing central nervous system and is co-expressed with Ret in several locations, including sympathetic, sensory, and parasympathetic ganglia, as well as in the ureteric buds of the developing kidneys. Pull-down assays using the Dok-6 phosphotyrosine binding (PTB) domain and GDNF-activated Ret indicate that Dok-6 binds to the phosphorylated Ret Tyr(1062) residue. Moreover, ligand activation of Ret resulted in phosphorylation of tyrosine residue(s) located within the unique C terminus of Dok-6 predominantly through a Src-dependent mechanism, indicating that Dok-6 is a substrate of the Ret-Src signaling pathway. (Crowder RJ, Enomoto H, Yang M, Johnson EM Jr, Milbrandt J. Dok-6, a Novel p62 Dok family member, promotes Ret-mediated neurite outgrowth. J Biol Chem. 2004 Oct 1;279(40):42072-81. Epub 2004 Jul 30. It is also known as DOK6, HsT3226, MGC20785 or IRS. 0.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6PKX4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 220164
Name Human DOK6 (aa 232-269) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Docking protein 6; DOK5L; Dok6; Dok-6; Downstream of tyrosine kinase 6; downstream of tyrosine kinases 6; HsT3226; RGD1564376
Common Name DOK6
Gene Symbol Dok6
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EQHERLMLEMEQKARLQTSLTEPMTLSKSISLPRSAYW
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.