missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DNAJC9 (aa 156-245) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57199 (PA5-57199. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
DjC9 (DnaJ subfamily C member 9) belongs to the Hsp40 (heat shock protein 40) family of proteins that function as co-chaperones to Hsp70.Hsp40 proteins are classified into 3 main subfamilies (A, B, and C). The families are defined by the presence of particular domains which include a highly conserved alpha-helical N-terminal domain termed the J-domain, a glycine/phenylalanine-rich region, a cysteine-rich region, and a C-terminal beta-sheet containing domain. DjC9 is part of subfamily C that bears only the J-domain. DjC9 has been shown to interact with HSP70 through its J domain and activate its ATPase activity.
Specifications
Specifications
| Accession Number | Q8WXX5 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 23234 |
| Name | Human DNAJC9 (aa 156-245) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 5330419I01Rik; AU020082; DnaJ (Hsp40) homolog, subfamily C, member 9; DnaJ heat shock protein family (Hsp40) member C9; dnaJ homolog subfamily C member 9; DnaJ protein SB73; Dnajc9; DnaJ-like factor; HDJC9; JDD1; SB73 |
| Common Name | DNAJC9 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Del en indholdskorrektion