missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DNAJC13 (aa 132-223) Control Fragment Recombinant Protein

Product Code. 30201032
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201032

Brand: Invitrogen™ RP96342

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57872 (PA5-57872. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

DNAJC13, also known as receptor-mediated endocytosis 8 (RME8), is the human homolog to a DnaJ domain-containing protein originally identified in a screen for endocytic defects in C. elegans. It is thought to be a co-chaperone of Hsc70 which regulates protein conformation at membrane sites and plays a role in intracellular trafficking, co-localizing with markers of the endosomal system. Recent experiments have indicated that the DNAJC13 protein is involved in membrane trafficking through early endosomes but not through degradative organelles. DNAJC13 has been also been shown to regulate the intracellular trafficking of the epidermal growth factor receptor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75165
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23317
Name Human DNAJC13 (aa 132-223) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias D030002L11Rik; DnaJ (Hsp40) homolog, subfamily C, member 13; DnaJ domain-containing protein RME-8; DnaJ heat shock protein family (Hsp40) member C13; dnaJ homolog subfamily C member 13; DNAJC13; Gm1124; j-domain containing protein, required for endocytosis; KIAA0678; mKIAA0678; PARK21; receptor mediated endocytosis RME-8; required for receptor-mediated endocytosis 8; Rme8; RME-8
Common Name DNAJC13
Gene Symbol DNAJC13
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VTPGGFDQINPATNRVLCSYDYRNIEGFVDLSDYQGGFCILYGGFSRLHLFASEQREEIIKSAIDHAGNYIGISLRIRKEPLEFEQYLNLRF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.